This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other NAPRT SIRNA Assays ›
Gene Symbol:
Naprt
Gene Name:
nicotinate phosphoribosyltransferase
Gene Aliases:
9130210N20Rik, Naprt1
Chromosome Location:
Chr.15: 75890964 - 75894481 on Build GRCm38
UniGene ID:
Mm.288368
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 170915

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Naprt
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172607.3 NP_766195.2 4 789
GenBank mRNA AK033661.1 - 4 792
AK049157.1 - 4 538
AK159127.1 - 4 588
AK170518.1 - 4 588
AY214331.1 - 4 792
BC058800.1 - 4 559 6476979

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

response to oxidative stress
NAD biosynthetic process
nicotinate nucleotide salvage
pyridine nucleotide biosynthetic process
nicotinate-nucleotide diphosphorylase (carboxylating) activity
nicotinate phosphoribosyltransferase activity
transferase activity
ligase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0