This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other BC030867 SIRNA Assays ›
Gene Symbol:
BC030867
Gene Name:
cDNA sequence BC030867
Gene Aliases:
-
Chromosome Location:
Chr.11: 102248865 - 102265183 on Build GRCm38
UniGene ID:
Mm.247335
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 170562

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: BC030867
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153544.3 NP_705772.3 5 1520
GenBank mRNA AK029811.1 - 5 1518
BC030867.1 - 5 1503
BC085317.1 - 5 1437
BC108338.1 - 5 1501 5034745

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001017988, XM_006247329, AY623029, AY623030.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0