This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CNST SIRNA Assays ›
Gene Symbol:
Cnst
Gene Name:
consortin, connexin sorting protein
Gene Aliases:
9630058J23Rik
Chromosome Location:
Chr.1: 179546349 - 179627473 on Build GRCm38
UniGene ID:
Mm.87662
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 170232

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cnst
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146105.3 NP_666217.2 2 504
GenBank mRNA AK036332.1 - 2 536
AK040666.1 - 2 527
AK040899.1 - 2 535
AK087361.1 - 2 522
AK146681.1 - 2 594
BC054802.1 - 2 504 6837803

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of phosphatase activity
positive regulation of Golgi to plasma membrane protein transport
protein binding
phosphatase binding
connexin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-9hb2s:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header