This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other APH1A SIRNA Assays ›
Gene Symbol:
Aph1a
Gene Name:
anterior pharynx defective 1a homolog (C. elegans)
Gene Aliases:
6530402N02Rik, APH-1a
Chromosome Location:
Chr.3: 95893907 - 95898592 on Build GRCm38
UniGene ID:
Mm.306954
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 170230

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Aph1a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146104.3 NP_666216.1 6 2500
NM_146134.2 NP_666246.1 7 2167
GenBank mRNA AK148400.1 - 6 2453
AK190617.1 - 1 252
BC012406.1 - 6 2425 3483214

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

metanephros development
proteolysis
membrane protein ectodomain proteolysis
Notch signaling pathway
Notch receptor processing
protein processing
membrane protein intracellular domain proteolysis
amyloid precursor protein catabolic process
positive regulation of catalytic activity
endopeptidase activity
protein binding
peptidase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0