This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GCA SIRNA Assays ›
Gene Symbol:
Gca
Gene Name:
grancalcin
Gene Aliases:
5133401E04Rik, AI573844
Chromosome Location:
Chr.2: 62664327 - 62694109 on Build GRCm38
UniGene ID:
Mm.219877
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 169883

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gca
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145523.3 NP_663498.1 2 276
GenBank mRNA AB088388.1 - 2 170
AF518325.1 - 2 260
AK033740.1 - 2 309
AK046347.1 - 2 318
AK049317.1 - 2 277
AK050540.1 - 2 318
AK078915.1 - 2 277
BC021450.1 - 2 270 5044040

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

proteolysis
calcium-dependent cysteine-type endopeptidase activity
calcium ion binding
protein homodimerization activity
metal ion binding
protein heterodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0