This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other BC005624 SIRNA Assays ›
Gene Symbol:
BC005624
Gene Name:
cDNA sequence BC005624
Gene Aliases:
-
Chromosome Location:
Chr.2: 30972833 - 30981941 on Build GRCm38
UniGene ID:
Mm.235238
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 169526

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: BC005624
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144885.2 NP_659134.1 3 260
GenBank mRNA AK158706.1 - 225
AK163554.1 - 233
AK168109.1 - 3 180
BC005624.1 - 3 205 3962182
BY333721.1 - 3 261

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001025003, XM_006233915, XM_008761665, BC091189.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-655dcf6cc8-78hlp:80/100.66.128.142:80.
git-commit: 0d764ad91c70359c551b4d51a80efc6847a0ee54
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0