This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other 4933431E20RIK SIRNA Assays ›
Gene Symbol:
4933431E20Rik
Gene Name:
RIKEN cDNA 4933431E20 gene
Gene Aliases:
-
Chromosome Location:
Chr.3: 107888850 - 107896213 on Build GRCm38
UniGene ID:
Mm.270500
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 168584

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: 4933431E20Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK030251.1 - 4 2335
AK036227.1 - 3 2286
AK147824.1 - 4 2310
BC060136.1 - 3 2713
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA NM_177858.1 - 2286 Chr. - - [-] on Build

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-fd98957d6-rbs6c:80/100.66.128.142:80.
git-commit: 278a916a254c4ca07f9ccbb2b9efa8cd7578b5be
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0