This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GM13103 SIRNA Assays ›
Gene Symbol:
Gm13103
Gene Name:
predicted gene 13103
Gene Aliases:
E330017N17, OTTMUSG00000010433
Chromosome Location:
Chr.4: 143846497 - 143853637 on Build GRCm38
UniGene ID:
Mm.252206
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 168165

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gm13103
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177571.3 NP_808239.2 4 1315
GenBank mRNA AK054347.1 - 4 1317
BB555205.2 - 1 114
BC096438.1 - 4 1312 30481183
DV662991.1 - 2 646

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0