This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other APPL2 SIRNA Assays ›
Gene Symbol:
Appl2
Gene Name:
adaptor protein, phosphotyrosine interaction, PH domain and leucine zipper containing 2
Gene Aliases:
Dip3b
Chromosome Location:
Chr.10: 83600033 - 83648854 on Build GRCm38
UniGene ID:
Mm.282985
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 167131

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Appl2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145220.2 NP_660255.1 8 573
GenBank mRNA AK154525.1 - 8 597
AK160075.1 - 8 552
AK160105.1 - 8 549
AK161003.1 - 8 580
AK168403.1 - 8 534
AK168748.1 - 8 529
AK171512.1 - 8 529
AY113706.1 - 8 575
BC048906.1 - 8 511 6475576

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cell cycle
cell proliferation
Rab GTPase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0