This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TAS2R131 SIRNA Assays ›
Gene Symbol:
Tas2r131
Gene Name:
taste receptor, type 2, member 131
Gene Aliases:
T2R31, Tas2r31, mGR31, mt2r61
Chromosome Location:
Chr.6: 132956912 - 132957844 on Build GRCm38
UniGene ID:
Mm.377917
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 166554

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tas2r131
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207030.1 NP_996913.1 1 902
GenBank mRNA BC145798.1 - 1 977 40131101

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0