siRNA Details
Gene Symbol: Tas2r131 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207030.1 | NP_996913.1 | 1 | 619 | ||
GenBank mRNA | BC145798.1 | - | 1 | 694 | 40131101 |

Gene Symbol: Tas2r131 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207030.1 | NP_996913.1 | 1 | 619 | ||
GenBank mRNA | BC145798.1 | - | 1 | 694 | 40131101 |