This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other WDTC1 SIRNA Assays ›
Gene Symbol:
Wdtc1
Gene Name:
WD and tetratricopeptide repeats 1
Gene Aliases:
Gm695, adipose, adp
Chromosome Location:
Chr.4: 133292466 - 133353476 on Build GRCm38
UniGene ID:
Mm.226576
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 166499

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Wdtc1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199306.4 NP_955010.1 7 943
GenBank mRNA AK131700.1 - 9 1103
AK184372.1 - 1 47
BC048824.1 - 7 737 4188979
BC057107.1 - 6 586 5697877

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001107908, XM_008764134, XM_008764135.

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
in utero embryonic development
glucose metabolic process
regulation of cell size
cellular response to insulin stimulus
multicellular organism growth
negative regulation of catalytic activity
negative regulation of fatty acid biosynthetic process
cellular chemical homeostasis
enzyme inhibitor activity
histone binding
histone deacetylase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0