This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other NKIRAS2 SIRNA Assays ›
Gene Symbol:
Nkiras2
Gene Name:
NFKB inhibitor interacting Ras-like protein 2
Gene Aliases:
2410003M04Rik, 4930527H08Rik, D630018G21Rik, KBRAS2
Chromosome Location:
Chr.11: 100622955 - 100627602 on Build GRCm38
UniGene ID:
Mm.274734
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 166097

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Nkiras2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028024.2 NP_082300.1 4 711
GenBank mRNA AK010371.1 - 4 711
AK015918.1 - 716
AK052645.1 - 4 711
AK083475.1 - 4 754
AK149696.1 - 4 855
AK169878.1 - 4 736
AK208908.1 - 1 29
BC013469.1 - 3 664 4219892
BC024398.1 - 3 654 5003001

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

signal transduction
small GTPase mediated signal transduction
nucleotide binding
GTP binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0