This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other UFD1L SIRNA Assays ›
Gene Symbol:
UFD1L
Gene Name:
ubiquitin fusion degradation 1 like (yeast)
Gene Aliases:
UFD1
Chromosome Location:
Chr.22: 19449941 - 19479215 on Build GRCh38
UniGene ID:
Hs.474213
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 16573

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: UFD1L
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001035247.2 NP_001030324.2 2 248
NM_005659.6 NP_005650.2 2 248
XM_011530361.2 XP_011528663.1 2 374
GenBank mRNA AF141201.1 - 1 118
AJ239058.1 - 1 118
AK225877.1 - 2 205
AK304734.1 - 2 218
AK315998.1 - 2 199
AY101594.1 - 2 175
BC001049.2 - 2 195 3507963
BC005087.1 - 2 196 3830774
BM761958.1 - 2 220
CB128364.1 - 2 248
CR456607.1 - 2 132
DB496066.1 - 2 232
U64444.1 - 2 196

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_053418, CK599711, BC161818, AF234601.
This siRNA also targets the following transcripts in Mouse: NM_011672, AK010461, AK147138, BC006630, AK088823, and others.

Gene Ontology Categories:

Function(s) Process(es)

skeletal system development
ubiquitin-dependent protein catabolic process
retrograde protein transport, ER to cytosol
error-free translesion synthesis
ER-associated misfolded protein catabolic process
thiol-dependent ubiquitin-specific protease activity
receptor binding
protein binding
protein complex binding
K48-linked polyubiquitin binding
ATPase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0