This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RPP40 SIRNA Assays ›
Gene Symbol:
Rpp40
Gene Name:
ribonuclease P 40 subunit
Gene Aliases:
D8Bwg1265e, Rnasep1
Chromosome Location:
Chr.13: 35895104 - 35911565 on Build GRCm38
UniGene ID:
Mm.35545
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 165038

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rpp40
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145938.4 NP_666050.3 7, 8 917
GenBank mRNA AK048810.1 - 917
BC024607.1 - 7, 8 862 4236826
BC037666.1 - 5, 6 3060

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

tRNA 5'-leader removal
tRNA processing
RNA phosphodiester bond hydrolysis, endonucleolytic
ribonuclease P activity
hydrolase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0