This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other WDFY2 SIRNA Assays ›
Gene Symbol:
Wdfy2
Gene Name:
WD repeat and FYVE domain containing 2
Gene Aliases:
5830485M08, B130024L21Rik, ProF, ZFYVE22
Chromosome Location:
Chr.14: 62837690 - 62961509 on Build GRCm38
UniGene ID:
Mm.1960
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 164109

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Wdfy2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175546.4 NP_780755.2 9 1136
GenBank mRNA AK045072.1 - 8 867
AK086275.1 - 9 1149
AK151817.1 - 9 1092
BC116632.1 - 9 1082 40056912

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_052950, AK058149, AK311177, AF411978, BC014004.
This siRNA also targets the following transcripts in Rat: NM_001107269, XM_008770751, XM_006252177, XM_008770752.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0