This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SLCO5A1 SIRNA Assays ›
Gene Symbol:
Slco5a1
Gene Name:
solute carrier organic anion transporter family, member 5A1
Gene Aliases:
4732454B15, A630033C23Rik, AA407151
Chromosome Location:
Chr.1: 12866549 - 12992932 on Build GRCm38
UniGene ID:
Mm.24481
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 163974

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Slco5a1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172841.2 NP_766429.2 1 962
GenBank mRNA AK028758.1 - 2 864
AK041736.1 - 1 963
AK048882.1 - 1 887
BC117943.1 - 1 741 40097907
BC117944.1 - 1 741

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001107898, XM_006237743, XM_008763495.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0