This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PRR3 SIRNA Assays ›
Gene Symbol:
Prr3
Gene Name:
proline-rich polypeptide 3
Gene Aliases:
4930540G07Rik, Cat56
Chromosome Location:
Chr.17: 35972539 - 35979825 on Build GRCm38
UniGene ID:
Mm.40036
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 163676

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Prr3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282012.1 NP_001268941.1 4 1515
NM_001282013.1 NP_001268942.1 6 1520
NM_001282024.1 NP_001268953.1 5 1614
NM_145487.3 NP_663462.2 4 1543
GenBank mRNA AJ504719.1 - 4 2313
BC016636.1 - 4 1461 2650633

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
poly(A) RNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0