This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SEC62 SIRNA Assays ›
Gene Symbol:
Sec62
Gene Name:
SEC62 homolog (S. cerevisiae)
Gene Aliases:
3100002M17Rik, AI844545, AW545092, Dtrp1, HTP1, Tloc1
Chromosome Location:
Chr.3: 30792774 - 30821263 on Build GRCm38
UniGene ID:
Mm.26017
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 163120

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Sec62
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027016.2 NP_081292.1 2 546
GenBank mRNA AK013922.1 - 2 361
AK088093.1 - 2 95
AK090078.1 - 2 96
BC017162.1 - 2 547
BC048532.1 - 2 96
BC053504.1 - 2 83
BC067202.1 - 2 81 6332701
BC071243.1 - 2 96

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

posttranslational protein targeting to membrane
transport
protein transport
molecular_function
protein transporter activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0