This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other NTS SIRNA Assays ›
Gene Symbol:
Nts
Gene Name:
neurotensin
Gene Aliases:
5033428E16Rik, NMN-125, NN, NT, NT/N, NTS1
Chromosome Location:
Chr.10: 102481756 - 102490418 on Build GRCm38
UniGene ID:
Mm.64201
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 162853

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Nts
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024435.2 NP_077755.1 3 288
GenBank mRNA AF304160.1 - 3 291
AK017202.1 - 3 305
AK017212.1 - 3 304
BC043024.1 - 3 288 5691298

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

visual learning
regulation of blood vessel size
neuropeptide hormone activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0