This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CSF3 SIRNA Assays ›
Gene Symbol:
Csf3
Gene Name:
colony stimulating factor 3 (granulocyte)
Gene Aliases:
Csfg, G-CSF, MGI-IG
Chromosome Location:
Chr.11: 98701095 - 98704836 on Build GRCm38
UniGene ID:
Mm.1238
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 161362

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Csf3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009971.1 NP_034101.1 5 801
GenBank mRNA BC120759.1 - 5 754 40129682
BC120761.1 - 5 754 40129684
M13926.1 - 5 801

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

immune response
positive regulation of cell proliferation
positive regulation of phosphatidylinositol 3-kinase signaling
positive regulation of actin filament polymerization
granulocyte differentiation
positive regulation of protein binding
positive regulation of peptidyl-serine phosphorylation
positive regulation of transcription factor import into nucleus
positive regulation of myeloid cell differentiation
positive regulation of transcription from RNA polymerase II promoter
positive regulation of peptidyl-tyrosine phosphorylation
positive regulation of protein kinase B signaling
cellular response to cytokine stimulus
negative regulation of neuron death
positive regulation of actin cytoskeleton reorganization
cytokine activity
growth factor activity
enzyme binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0