This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CRYBA2 SIRNA Assays ›
Gene Symbol:
CRYBA2
Gene Name:
crystallin beta A2
Gene Aliases:
CTRCT42
Chromosome Location:
Chr.2: 218990190 - 218993405 on Build GRCh38
UniGene ID:
Hs.415790
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 16086

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CRYBA2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_057093.1 NP_476434.1 3 651
NM_057094.1 NP_476435.1 4 457
GenBank mRNA AF166331.1 - 4 451
BC006285.2 - 3 651 3960073
BF726935.1 - 4 465
BT007447.1 - 3 422

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

lens development in camera-type eye
biological_process
molecular_function
structural constituent of eye lens
protein binding
protein homodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0