This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GPR162 SIRNA Assays ›
Gene Symbol:
Gpr162
Gene Name:
G protein-coupled receptor 162
Gene Aliases:
A-2, BC031437, Grca
Chromosome Location:
Chr.6: 124858444 - 124864219 on Build GRCm38
UniGene ID:
Mm.2514
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 159253

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gpr162
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013533.3 NP_038561.1 5 2125
GenBank mRNA AK134941.1 - 5 1939
AK144354.1 - 4 1894
AK144543.1 - 5 1912
AK158847.1 - 4 3017
AK171145.1 - 6 2309
BC016413.1 - 4 1555
BC031437.1 - 5 2125 4527410

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001108646, XM_006237371.

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
G-protein coupled receptor signaling pathway
biological_process
molecular_function
signal transducer activity
G-protein coupled receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0