This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other HOXD3 SIRNA Assays ›
Gene Symbol:
Hoxd3
Gene Name:
homeobox D3
Gene Aliases:
Hox-4.1, Hox-5.5
Chromosome Location:
Chr.2: 74711993 - 74748271 on Build GRCm38
UniGene ID:
Mm.430708
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 159027

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Hoxd3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010468.2 NP_034598.2 3 2177
GenBank mRNA AK133705.1 - 3 2045
AK138525.1 - 3 2178
AK140904.1 - 1 1825
U56400.1 - 2 1853
X73573.1 - 2 1833

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001271038, XM_008761880, XM_008761881.

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
cell-matrix adhesion
Notch signaling pathway
multicellular organism development
anterior/posterior pattern specification
positive regulation of gene expression
glossopharyngeal nerve morphogenesis
thyroid gland development
positive regulation of neuron differentiation
embryonic skeletal system morphogenesis
cartilage development
DNA binding
transcription factor activity, sequence-specific DNA binding
sequence-specific DNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0