This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other HIPK3 SIRNA Assays ›
Gene Symbol:
Hipk3
Gene Name:
homeodomain interacting protein kinase 3
Gene Aliases:
DYRK6, FIST3, PKY, mir-1902
Chromosome Location:
Chr.2: 104426481 - 104494489 on Build GRCm38
UniGene ID:
Mm.257925
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 158964

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Hipk3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145824.1 NP_001139296.1 13 3182
NM_010434.2 NP_034564.2 13 3185
GenBank mRNA AF077660.1 - 13 2939
AF170305.1 - 12 2649
AF305238.1 - 12 2647
AK140130.1 - 13 3288
BC131663.1 - 13 2817
BC131664.1 - 13 2813 40110751
X97572.1 - 3 403

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_031787, XM_008762072, XM_006234658, AF036959, BC100068.

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription, DNA-templated
protein phosphorylation
apoptotic process
mRNA transcription
phosphorylation
peptidyl-serine phosphorylation
peptidyl-threonine phosphorylation
negative regulation of apoptotic process
positive regulation of DNA binding
negative regulation of JUN kinase activity
nucleotide binding
DNA binding
transcription corepressor activity
protein kinase activity
protein serine/threonine kinase activity
protein binding
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0