This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other AES SIRNA Assays ›
Gene Symbol:
Aes
Gene Name:
amino-terminal enhancer of split
Gene Aliases:
AL024115, Esp1, Grg, Grg-5, Grg5, Tle5
Chromosome Location:
Chr.10: 81559444 - 81566371 on Build GRCm38
UniGene ID:
Mm.180013
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 158830

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Aes
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001276288.1 NP_001263217.1 7 737
NM_010347.4 NP_034477.1 7 740
GenBank mRNA AK019397.1 - 1 163
AK131845.1 - 7 617
AK149015.1 - 4 1408
AK158714.1 - 7 625
AK189802.1 - 1 113
AK207707.1 - 1 112
BC132217.1 - 7 628 40130494
BC132219.1 - 7 628 40130496
BC145299.1 - 7 625 9053853
L12140.1 - 7 581
X73359.1 - 6 691
X73361.1 - 7 640

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_019220, BC101855, L14462.

Panther Classification:

Molecular Function -

transcription cofactor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
skeletal system development
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
negative regulation of gene expression
Wnt signaling pathway
cellular response to extracellular stimulus
negative regulation of protein binding
regulation of growth
negative regulation of transcription, DNA-templated
negative regulation of response to cytokine stimulus
response to interleukin-1
negative regulation of canonical Wnt signaling pathway
positive regulation of anoikis
transcription corepressor activity
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0