This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FEM1A SIRNA Assays ›
Gene Symbol:
Fem1a
Gene Name:
feminization 1 homolog a (C. elegans)
Gene Aliases:
AW611390, Fem1aa
Chromosome Location:
Chr.17: 56256793 - 56263608 on Build GRCm38
UniGene ID:
Mm.290813
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 158506

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fem1a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010192.4 NP_034322.3 1 3009
GenBank mRNA AK034412.1 - 1 2984
AK075860.1 - 1 1133
AK080978.1 - 1 2982
AK129000.1 - 1 2992
AK147879.1 - 1 2994
AK155363.1 - 1 2993
BC009161.1 - 1 2977 2811079
BC054382.1 - 1 2980 6469158
BC110669.1 - 1 2963 5042239

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of MAP kinase activity
negative regulation of inflammatory response
transcription factor binding
EP4 subtype prostaglandin E2 receptor binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0