This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other EYA4 SIRNA Assays ›
Gene Symbol:
Eya4
Gene Name:
eyes absent 4 homolog (Drosophila)
Gene Aliases:
B130023L16Rik
Chromosome Location:
Chr.10: 23102962 - 23350856 on Build GRCm38
UniGene ID:
Mm.332128
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 158449

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Eya4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010167.4 NP_034297.2 9 878
GenBank mRNA AK041625.1 - 9 879
BC120898.1 - 9 862 40061597
BC120899.1 - 9 862 40061599
Y17115.1 - 10 989

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

DNA repair
transcription, DNA-templated
regulation of transcription, DNA-templated
cellular response to DNA damage stimulus
multicellular organism development
sensory perception of sound
chromatin modification
middle ear morphogenesis
molecular_function
phosphoprotein phosphatase activity
protein tyrosine phosphatase activity
hydrolase activity
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0