This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PPM1J SIRNA Assays ›
Gene Symbol:
PPM1J
Gene Name:
protein phosphatase, Mg2+/Mn2+ dependent 1J
Gene Aliases:
PP2CZ, PP2Czeta, PPP2CZ
Chromosome Location:
Chr.1: 112709994 - 112715328 on Build GRCh38
UniGene ID:
Hs.655231
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 15797

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: PPM1J
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005167.5 NP_005158.5 4 848
GenBank mRNA AB451327.1 - 3 203
AB451471.1 - 3 203
AK093270.1 - 4 828
AL133644.1 - 5 1612
BC011803.2 - 4 772
BC064628.1 - 4 778
BC073828.1 - 4 840 5244363

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein dephosphorylation
protein serine/threonine phosphatase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0