This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FST SIRNA Assays ›
Gene Symbol:
Fst
Gene Name:
follistatin
Gene Aliases:
AL033346, FS
Chromosome Location:
Chr.13: 114452262 - 114458951 on Build GRCm38
UniGene ID:
Mm.4913
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 157599

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fst
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001301373.1 NP_001288302.1 2 586
NM_001301375.1 NP_001288304.1 2 586
NM_008046.3 NP_032072.1 2 586
GenBank mRNA AK079916.1 - 2 587
AK083556.1 - 2 365
BC144926.1 - 2 410 9053474
BC145945.1 - 2 410 40131248
X83377.1 - 2 215
Z29532.1 - 2 221

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_012561, XM_006231953, XM_006231954.

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
skeletal system development
hematopoietic progenitor cell differentiation
gamete generation
pattern specification process
female gonad development
BMP signaling pathway
hair follicle morphogenesis
negative regulation of activin receptor signaling pathway
odontogenesis of dentin-containing tooth
keratinocyte proliferation
negative regulation of cell differentiation
positive regulation of hair follicle development
heparan sulfate proteoglycan binding
activin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7bff9b55f5-ncxmj:80/100.66.128.142:80.
git-commit: 5f36da1a8ad9bc9722227bfd86ebc827ae27d083
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0