This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MECOM SIRNA Assays ›
Gene Symbol:
Mecom
Gene Name:
MDS1 and EVI1 complex locus
Gene Aliases:
D630039M04Rik, Evi-1, Evi1, Jbo, Mds, Mds1, Mds1-Evi1, Prdm3, Znfpr1b1
Chromosome Location:
Chr.3: 29951296 - 30509783 on Build GRCm38
UniGene ID:
Mm.426829
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 157408

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Mecom
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007963.2 NP_031989.2 6 1301
GenBank mRNA AJ001482.1 - 4 457
AK033517.1 - 6 735
AK051098.1 - 6 1161
AK085548.1 - 6 1305
BC076620.1 - 6 1253
BC139762.1 - 4 419 40109903
JQ665270.1 - 4 449

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001106423, XM_006232249, XM_008760909, XM_008760910, XM_006232250, and others.

Gene Ontology Categories:

Function(s) Process(es)

in utero embryonic development
neutrophil homeostasis
inflammatory response
response to bacterium
post-embryonic development
stem cell population maintenance
forebrain development
embryonic forelimb morphogenesis
embryonic hindlimb morphogenesis
regulation of cell proliferation
negative regulation of programmed cell death
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
negative regulation of JNK cascade
regulation of cell cycle
pericardium development
renal system development
positive regulation of brown fat cell differentiation
DNA binding
transcription factor activity, sequence-specific DNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-fd98957d6-rbs6c:80/100.66.128.142:80.
git-commit: 278a916a254c4ca07f9ccbb2b9efa8cd7578b5be
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0