This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MSX3 SIRNA Assays ›
Gene Symbol:
Msx3
Gene Name:
msh homeobox 3
Gene Aliases:
AI323377
Chromosome Location:
Chr.7: 140046156 - 140049970 on Build GRCm38
UniGene ID:
Mm.4816
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 156383

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Msx3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010836.3 NP_034966.1 2 424
GenBank mRNA AB221640.1 - 2 335
AK012208.1 - 2 424
AK012556.1 - 2 493
BC051983.1 - 2 393
BC137577.1 - 2 423 8860596
BC137578.1 - 2 423 8860597
U62523.1 - 2 431

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
multicellular organism development
negative regulation of histone acetylation
DNA binding
protein binding
sequence-specific DNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0