This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MAN1A2 SIRNA Assays ›
Gene Symbol:
Man1a2
Gene Name:
mannosidase, alpha, class 1A, member 2
Gene Aliases:
AI428775, AI528764, AI854422, Man1b, PCR2
Chromosome Location:
Chr.3: 100562203 - 100685473 on Build GRCm38
UniGene ID:
Mm.271617
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 156239

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Man1a2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010763.2 NP_034893.1 13 3122
GenBank mRNA AK153336.1 - 13 3122
BC049121.1 - 13 2526 6813258
BC068192.1 - 13 2526 6813258
U03457.1 - 13 3100
U03458.1 - 13 3100

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

respiratory gaseous exchange
metabolic process
glycoprotein metabolic process
lung alveolus development
mannosyl-oligosaccharide 1,2-alpha-mannosidase activity
calcium ion binding
lytic transglycosylase activity
mannosidase activity
mannosyl-oligosaccharide mannosidase activity
galactosidase activity
glucosidase activity
trehalase activity
fucosidase activity
hexosaminidase activity
amylase activity
hydrolase activity
hydrolase activity, acting on glycosyl bonds
dextrin alpha-glucosidase activity
starch alpha-glucosidase activity
beta-glucanase activity
beta-6-sulfate-N-acetylglucosaminidase activity
glucan endo-1,4-beta-glucosidase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0