This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZFP324 SIRNA Assays ›
Gene Symbol:
Zfp324
Gene Name:
zinc finger protein 324
Gene Aliases:
A930002M05, D430030K24Rik, ZF5128
Chromosome Location:
Chr.7: 12965811 - 12974244 on Build GRCm38
UniGene ID:
Mm.133086
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 154611

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Zfp324
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178732.3 NP_848847.3 4 2595
GenBank mRNA AK051488.1 - 4 2618
AK052251.1 - 3 2488
AK054407.1 - 4 2640
AK085053.1 - 3 2491
BC053093.1 - 4 2595 6400158

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0