This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PARP12 SIRNA Assays ›
Gene Symbol:
Parp12
Gene Name:
poly (ADP-ribose) polymerase family, member 12
Gene Aliases:
9930021O16, AA409132, AA536654, ARTD12, PARP-12, Zc3hdc1
Chromosome Location:
Chr.6: 39086412 - 39118349 on Build GRCm38
UniGene ID:
Mm.268462
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 153398

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Parp12
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172893.3 NP_766481.2 12 2752
GenBank mRNA AK036886.1 - 12 2756
AK053205.1 - 12 2653
AK081528.1 - 12 2658
AK156623.1 - 12 2621
AK162200.1 - 12 2324
BC024579.1 - 7 1401
BC028906.1 - 4 1112
BC048927.1 - 8 1603

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
NAD+ ADP-ribosyltransferase activity
transferase activity
poly(A) RNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0