This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SATB1 SIRNA Assays ›
Gene Symbol:
Satb1
Gene Name:
special AT-rich sequence binding protein 1
Gene Aliases:
2610306G12Rik, AW413156
Chromosome Location:
Chr.17: 51736187 - 51833497 on Build GRCm38
UniGene ID:
Mm.311655
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 150686

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Satb1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163630.1 NP_001157102.1 11 2784
NM_001163631.1 NP_001157103.1 11 2652
NM_001163632.1 NP_001157104.1 11 2625
NM_009122.2 NP_033148.2 11 2675
GenBank mRNA AK037740.1 - 11 2676
AK088204.1 - 1 260
AK088563.1 - 11 2651
AK134040.1 - 11 2654
AK137875.1 - 369
AK158518.1 - 11 2785
BC011132.1 - 11 2535 4164993
U05252.1 - 11 2661

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
chromatin organization
chromatin remodeling
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
epidermis development
chromatin modification
histone methylation
T cell activation
CD4-positive, alpha-beta T cell differentiation
CD8-positive, alpha-beta T cell differentiation
activated T cell proliferation
reflex
RNA polymerase II regulatory region sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0