siRNA Details
Gene Symbol: LINC00336 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027908.1 | 2 | 965 | |||
GenBank mRNA | AK125740.1 | - | 2 | 965 |

Gene Symbol: LINC00336 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027908.1 | 2 | 965 | |||
GenBank mRNA | AK125740.1 | - | 2 | 965 |