siRNA Details
Gene Symbol: CLEC9A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207345.3 | NP_997228.1 | 7 | 1054 | ||
GenBank mRNA | AY358265.1 | - | 7 | 1054 | ||
EU339276.1 | - | 4 | 441 |

Gene Symbol: CLEC9A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207345.3 | NP_997228.1 | 7 | 1054 | ||
GenBank mRNA | AY358265.1 | - | 7 | 1054 | ||
EU339276.1 | - | 4 | 441 |