This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SPATA19 SIRNA Assays ›
Gene Symbol:
SPATA19
Gene Name:
spermatogenesis associated 19
Gene Aliases:
CT132, SPAS1, spergen1
Chromosome Location:
Chr.11: 133835930 - 133845538 on Build GRCh38
UniGene ID:
Hs.97541
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 149304

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: SPATA19
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291992.1 NP_001278921.1 4 427
NM_174927.2 NP_777587.1 4 427
GenBank mRNA AF521713.1 - 4 363
AK098717.1 - 4 386
BC058039.1 - 4 394 6619434
BX096445.1 - 4 427

Back To Top

More Information


Gene Ontology Categories:

Function(s)

multicellular organism development
spermatogenesis
cell differentiation

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0