This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZNF614 SIRNA Assays ›
Gene Symbol:
ZNF614
Gene Name:
zinc finger protein 614
Gene Aliases:
-
Chromosome Location:
Chr.19: 52013324 - 52028427 on Build GRCh38
UniGene ID:
Hs.292336
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 148912

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF614
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025040.3 NP_079316.2 5 2365
GenBank mRNA AK097156.1 - 5 2349
BC022246.1 - 1 1164
BC101392.1 - 5 2203 40026900
BC101393.2 - 5 2203 40026901
BC101394.1 - 5 2203 40026907
BC101395.1 - 5 2203 40026909
BG286230.1 - 1 115

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
biological_process
molecular_function
DNA binding
protein binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0