This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KIAA1456 SIRNA Assays ›
Gene Symbol:
KIAA1456
Gene Name:
KIAA1456
Gene Aliases:
C8orf79, TRM9L
Chromosome Location:
Chr.8: 12945674 - 13029777 on Build GRCh38
UniGene ID:
Hs.202521
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
148788
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KIAA1456
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001099677.1
NP_001093147.1
2
669
NM_020844.2
NP_065895.2
5
959
XM_005273584.4
XP_005273641.1
5
970
XM_005273585.4
XP_005273642.1
6
981
XM_005273586.4
XP_005273643.1
6
1015
XM_005273587.4
XP_005273644.1
5
1203
XM_005273588.4
XP_005273645.1
6
1127
XM_005273590.4
XP_005273647.1
4
659
XM_005273591.4
XP_005273648.1
5
1143
XM_011544596.2
XP_011542898.1
5
1430
XM_011544597.2
XP_011542899.1
5
782
XM_011544598.2
XP_011542900.1
5
1051
XM_011544600.2
XP_011542902.1
5
789
XM_011544601.2
XP_011542903.1
5
912
XM_017013705.1
XP_016869194.1
5
820
XM_017013706.1
XP_016869195.1
5
1041
XM_017013707.1
XP_016869196.1
6
1186
XM_017013708.1
XP_016869197.1
5
1285
XM_017013709.1
XP_016869198.1
6
1170
XM_017013710.1
XP_016869199.1
7
1129
XM_017013711.1
XP_016869200.1
6
964
XM_017013712.1
XP_016869201.1
6
1341
XM_017013713.1
XP_016869202.1
5
1085
XM_017013714.1
XP_016869203.1
4
887
XM_017013715.1
XP_016869204.1
5
1760
GenBank mRNA
AB040889.1
-
2
733
AK307825.1
-
2
984
BC016633.2
-
2
312
DR001645.1
-
2
440
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
tRNA wobble uridine modification
tRNA modification
tRNA methylation
oxidation-reduction process
tRNA binding
tRNA methyltransferase activity
ferrous iron binding
tRNA (uracil) methyltransferase activity
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, 2-oxoglutarate as one donor, and incorporation of one atom each of oxygen into both donors
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0