This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CWF19L1 SIRNA Assays ›
Gene Symbol:
CWF19L1
Gene Name:
CWF19-like 1, cell cycle control (S. pombe)
Gene Aliases:
C19L1, SCAR17, hDrn1
Chromosome Location:
Chr.10: 100232296 - 100267681 on Build GRCh38
UniGene ID:
Hs.215502
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 148714

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CWF19L1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001303404.1 NP_001290333.1 5 469
NM_001303405.1 NP_001290334.1 5 514
NM_001303406.1 NP_001290335.1 2 203
NM_018294.5 NP_060764.3 5 469
GenBank mRNA AK001860.1 - 5 415
AK225590.1 - 5 427
AK295303.1 - 2 173
AL832515.1 - 5 426
BC008746.2 - 5 410 3345869
DC299678.1 - 5 469
KU178692.1 - 5 381
KU178693.1 - 5 365

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function
catalytic activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0