This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other HYDIN SIRNA Assays ›
Gene Symbol:
HYDIN
Gene Name:
HYDIN, axonemal central pair apparatus protein
Gene Aliases:
CILD5, HYDIN1, HYDIN2, PPP1R31
Chromosome Location:
Chr.16: 70802084 - 71230722 on Build GRCh38
UniGene ID:
Hs.351914
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 148652

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: HYDIN
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001198542.1 NP_001185471.1 5, 6 640
NM_001198543.1 NP_001185472.1 5, 6 610
NM_001270974.2 NP_001257903.1 5, 6 708
NM_017558.4 NP_060028.2 5, 6 708
XM_006721206.2 XP_006721269.1 5, 6 602
XM_011523146.2 XP_011521448.1 5, 6 635
XM_011523147.1 XP_011521449.1 5, 6 602
XM_011523148.1 XP_011521450.1 5, 6 675
XM_011523151.1 XP_011521453.1 5, 6 634
XM_017023346.1 XP_016878835.1 5, 6 880
GenBank mRNA AK022933.1 - 5, 6 642
AK299016.1 - 5, 6 597
AK299348.1 - 5, 6 640
BC028351.1 - 5, 6 589

Back To Top

More Information


Gene Ontology Categories:

Function(s)

epithelial cell development
cilium movement
ventricular system development
trachea development
axonemal central apparatus assembly

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0