This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MGRN1 SIRNA Assays ›
Gene Symbol:
MGRN1
Gene Name:
mahogunin ring finger 1
Gene Aliases:
RNF156
Chromosome Location:
Chr.16: 4616493 - 4690974 on Build GRCh38
UniGene ID:
Hs.526494
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 148546

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: MGRN1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142289.2 NP_001135761.2 14 1579
NM_001142290.2 NP_001135762.1 15 1645
NM_001142291.2 NP_001135763.2 14 1579
NM_015246.3 NP_056061.1 15 1645
NR_102267.1 13 1469
XM_005255217.1 XP_005255274.1 15 1648
XM_005255218.1 XP_005255275.1 14 1582
XM_005255219.3 XP_005255276.1 15 1648
XM_005255220.3 XP_005255277.1 15 1645
XM_005255221.3 XP_005255278.1 14 1579
XM_011522441.1 XP_011520743.1 15 1506
XM_011522442.2 XP_011520744.1 7 960
XM_017023091.1 XP_016878580.1 15 1648
XM_017023092.1 XP_016878581.1 14 1582
XM_017023093.1 XP_016878582.1 15 1503
GenBank mRNA AB011116.1 - 15 1601
AK299052.1 - 13 1451
BC050389.1 - 15 1617 5755971
EF471397.2 - 14 1442
EF471398.2 - 14 1442

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
protein monoubiquitination
endosome to lysosome transport
negative regulation of cAMP-mediated signaling
negative regulation of G-protein coupled receptor protein signaling pathway
ubiquitin-protein transferase activity
protein binding
zinc ion binding
ligase activity
ubiquitin protein ligase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0