This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GSE1 SIRNA Assays ›
Gene Symbol:
GSE1
Gene Name:
Gse1 coiled-coil protein
Gene Aliases:
CRHSP24, KIAA0182
Chromosome Location:
Chr.16: 85169512 - 85676206 on Build GRCh38
UniGene ID:
Hs.461647
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 148446

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: GSE1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134473.2 NP_001127945.1 15 5962
NM_001278184.1 NP_001265113.1 15 6016
NM_014615.3 NP_055430.1 16 6235
XM_005255859.4 XP_005255916.3 17 8377
XM_005255860.3 XP_005255917.3 16 8270
XM_005255861.4 XP_005255918.3 16 8158
XM_005255863.4 XP_005255920.1 16 6127
XM_005255864.4 XP_005255921.1 16 6859
XM_005255865.4 XP_005255922.1 15 6640
XM_011522965.2 XP_011521267.1 16 6626
XM_011522966.2 XP_011521268.1 16 6093
XM_017023083.1 XP_016878572.1 16 6385
GenBank mRNA AK127344.1 - 5 4638
D80004.2 - 15 5889

Back To Top

More Information


Gene Ontology Categories:

Process(es)

protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header