This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZNF644 SIRNA Assays ›
Gene Symbol:
ZNF644
Gene Name:
zinc finger protein 644
Gene Aliases:
BM-005, MYP21, NatF, ZEP-2
Chromosome Location:
Chr.1: 90915298 - 91022269 on Build GRCh38
UniGene ID:
Hs.173001
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 148280

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF644
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016620.3 NP_057704.2 4 1039
NM_032186.4 NP_115562.3 4 1069
NM_201269.2 NP_958357.1 6 4713
XM_005271257.4 XP_005271314.1 8 5172
XM_005271260.4 XP_005271317.1 7 5149
XM_011542258.2 XP_011540560.1 7 4850
XM_011542259.2 XP_011540561.1 9 5526
XM_011542260.2 XP_011540562.1 9 5430
XM_011542261.2 XP_011540563.1 7 4771
XM_017002487.1 XP_016857976.1 8 4961
XM_017002488.1 XP_016857977.1 7 5049
XM_017002489.1 XP_016857978.1 6 5024
XM_017002490.1 XP_016857979.1 7 4836
XM_017002491.1 XP_016857980.1 6 4648
XM_017002492.1 XP_016857981.1 5 2059
XM_017002493.1 XP_016857982.1 5 1405
XM_017002494.1 XP_016857983.1 5 1182
GenBank mRNA AB033047.1 - 4 4296
AK001587.1 - 4 2478
AK023596.1 - 4 1652
BC031003.1 - 3 1100
BC050656.2 - 4 988
BC063683.1 - 5 4407
BC110841.1 - 4 887
BC132776.1 - 6 4528 40146798
BC132778.1 - 6 4528 40146800
BC150177.1 - 6 4668 40148078

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0