This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZNF570 SIRNA Assays ›
Gene Symbol:
ZNF570
Gene Name:
zinc finger protein 570
Gene Aliases:
-
Chromosome Location:
Chr.19: 37467465 - 37488652 on Build GRCh38
UniGene ID:
Hs.126962
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 148049

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF570
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001300993.2 NP_001287922.1 6 713
NM_001321991.1 NP_001308920.1 5 562
NM_001321992.1 NP_001308921.1 4 893
NM_001321993.1 NP_001308922.1 5 1036
NM_001321994.1 NP_001308923.1 4 952
NM_144694.3 NP_653295.1 5 977
XM_011526544.2 XP_011524846.1 5 560
XM_011526545.2 XP_011524847.1 4 464
GenBank mRNA AK055353.1 - 5 955
AK297561.1 - 4 654
AK308252.1 - 6 713
BC130425.1 - 5 860 40146456
BC130433.1 - 5 860 40146464

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
RNA polymerase II regulatory region sequence-specific DNA binding
transcription factor activity, sequence-specific DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header