This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CALCB SIRNA Assays ›
Gene Symbol:
CALCB
Gene Name:
calcitonin related polypeptide beta
Gene Aliases:
CALC2, CGRP-II, CGRP2
Chromosome Location:
Chr.11: 15073600 - 15078631 on Build GRCh38
UniGene ID:
Hs.534305
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 145616

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CALCB
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000728.3 NP_000719.1 5 822
GenBank mRNA AK054728.1 - 10 2044
BC092468.1 - 5 826 5313089
M28641.1 - 1 26
X02404.1 - 3 587

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cellular calcium ion homeostasis
signal transduction
mitophagy in response to mitochondrial depolarization
neuropeptide hormone activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0