This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GOLGA2 SIRNA Assays ›
Gene Symbol:
GOLGA2
Gene Name:
golgin A2
Gene Aliases:
GM130
Chromosome Location:
Chr.9: 128255829 - 128276007 on Build GRCh38
UniGene ID:
Hs.155827
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 145093

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: GOLGA2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004486.4 NP_004477.3 13 1005
XM_005251930.1 XP_005251987.1 14 1104
XM_005251931.1 XP_005251988.1 13 1023
XM_005251932.1 XP_005251989.1 12 935
GenBank mRNA AF248953.1 - 13 956
BC014188.2 - 1 22
BC069268.1 - 13 1005 6340972

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein glycosylation
ER to Golgi vesicle-mediated transport
microtubule nucleation
asymmetric cell division
negative regulation of autophagy
negative regulation of protein binding
COPII vesicle coating
spindle assembly
protein homotetramerization
centrosome organization
positive regulation of protein glycosylation
Golgi ribbon formation
Golgi disassembly
spindle assembly involved in meiosis
mitotic spindle assembly
cell-cell adhesion
protein binding
microtubule binding
protein kinase binding
syntaxin binding
importin-alpha family protein binding
cadherin binding involved in cell-cell adhesion

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0