This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FLT3LG SIRNA Assays ›
Gene Symbol:
FLT3LG
Gene Name:
fms related tyrosine kinase 3 ligand
Gene Aliases:
FL, FLT3L
Chromosome Location:
Chr.19: 49474172 - 49487038 on Build GRCh38
UniGene ID:
Hs.428
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 144682
Validated

Purification
Size
Availability Inventoried
Catalog # AM51331
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FLT3LG
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001204502.1 NP_001191431.1 5 426
NM_001204503.1 NP_001191432.1 4 395
NM_001278637.1 NP_001265566.1 5 454
NM_001278638.1 NP_001265567.1 5 443
NM_001459.3 NP_001450.2 5 426
XM_005258680.4 XP_005258737.3 4 416
XM_005258681.4 XP_005258738.3 4 416
XM_005258682.4 XP_005258739.3 4 417
XM_005258683.4 XP_005258740.3 4 416
XM_006723116.3 XP_006723179.2 4 417
XM_011526675.2 XP_011524977.1 5 462
XM_011526676.2 XP_011524978.1 5 427
XM_011526677.2 XP_011524979.1 4 416
XM_011526678.2 XP_011524980.1 5 462
XM_011526680.2 XP_011524982.1 4 416
XM_011526682.2 XP_011524984.1 4 433
XM_017026532.1 XP_016882021.1 4 417
XM_017026533.1 XP_016882022.1 4 352
XM_017026534.1 XP_016882023.1 4 325
XM_017026535.1 XP_016882024.1 5 449
GenBank mRNA AI792400.1 - 5 407
AK301136.1 - 4 387
BC006331.2 - 5 416
BC011914.2 - 5 416
BC028001.1 - 5 427 5198812
BC126293.1 - 5 383 8992009
BC136464.1 - 5 383 9020453
BC144129.1 - 5 383 9052649
DB490139.2 - 5 457
U03858.1 - 4 399
U04806.1 - 5 408

Back To Top

More Information


Matching Taqman Gene Expression Assay:


Hs00181740_m1

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of protein phosphorylation
signal transduction
positive regulation of cell proliferation
lymphocyte differentiation
regulation of myeloid dendritic cell activation
positive regulation of natural killer cell differentiation
embryonic hemopoiesis
positive regulation of myoblast differentiation
positive regulation of cell cycle
positive regulation of transcription from RNA polymerase II promoter
homeostasis of number of cells within a tissue
positive regulation of cell proliferation in bone marrow
negative regulation of apoptotic process in bone marrow
positive regulation of osteoclast proliferation
positive regulation of myoblast fusion
receptor binding
cytokine activity
receptor tyrosine kinase binding
protein homodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0